2,5-Dimethoxybenzhydrazide

Supplier: Matrix Scientific

003996-1G 003996-5G
101786-610EA 38.46 USD
101786-610 101786-612
2,5-Dimethoxybenzhydrazide
2,5-Dimethoxybenzhydrazide

Hazard code(s): Irritant

Formula: C₉H₁₂N₂O₃
Melting Pt: 89…91 °C
MDL Number: MFCD00017049
CAS Number: 17894-25-6

Order Now

Specification Test Results

Melting point 89-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR