3,4-Dimethoxy-β-methyl-β-nitrostyrene

Supplier: Matrix Scientific
Warning

003992-5G 003992-25G
101786-602EA 150.77 USD
101786-602 101786-604
3,4-Dimethoxy-β-methyl-β-nitrostyrene
3,4-Dimethoxy-β-methyl-β-nitrostyrene

Hazard code(s): Irritant

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 68…71 °C
MDL Number: MFCD00024815
CAS Number: 122-47-4
EINECS: 204-547-8

Order Now

Specification Test Results

Melting point 68-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR