(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid

003990-1G 003990-5G
101786-594EA 136.92 USD
101786-594 101786-596
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00016834
CAS Number: 4385-56-2
EINECS: 224-494-4

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR