4-Methyl-2-thiopheneboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid

003967-1G 003967-5G
101786-536EA 158.46 USD
101786-536 101786-538
4-Methyl-2-thiopheneboronic acid ≥97%
4-Methyl-2-thiopheneboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₇BO₂S
MW: 141.99 g/mol
MDL Number: MFCD01319040
CAS Number: 162607-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR