4-Chlorothioanisole ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide

003954-25G 003954-100G
101786-520EA 26.15 USD
101786-520 101786-522
4-Chlorothioanisole ≥99%
4-Chlorothioanisole

Hazard code(s): Stench

Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 168…169 °C
Melting Pt: 27…28 °C
Density: 1.2151 g/cm³ (19 °C)
MDL Number: MFCD00013643
CAS Number: 123-09-1
EINECS: 204-600-5

Order Now

Specification Test Results

Boiling point 168-169°C
Density 1.22

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR