Perfluoro-1-heptene ≥90%

Supplier: Matrix Scientific

003934-1G 003934-5G
101786-474EA 272.31 USD
101786-474 101786-476
Perfluoro-1-heptene ≥90%
Perfluoro-1-heptene

contains up to 10% perfluoroheptene-2


  • n20/D: 1.2782
  • Hazard code(s): Irritant, Refrigerate


Hazard code(s): Irritant, Refrigerate

Formula: C₇F₁₄
Boiling Pt: 80…82 °C
MDL Number: MFCD00013577
CAS Number: 355-63-5
EINECS: 206-590-8

Order Now

Specification Test Results

Boiling point 80-82°C
ND20 1.2782


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR