3-Fluorophenylhydrazinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: (3-Fluorphenyl)hydrazin HCl, (3-Fluorophenyl)hydrazine hydrochloride, 3-Fluorophenylhydrazine hydrochloride, 1-(3-Fluorophenyl)hydrazine hydrochloride

003925-5G 003925-25G
101786-466EA 23.08 USD
101786-466 101786-468
3-Fluorophenylhydrazinium chloride ≥97%
3-Fluorophenylhydrazinium chloride

Hazard code(s): Irritant-Harmful

Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 268 °C
MDL Number: MFCD00012934
CAS Number: 2924-16-5
EINECS: 220-886-4

Order Now

Specification Test Results

Melting point 268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR