2-Fluorophenylhydrazinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride

003924-25G 003924-100G
101786-462EA 35.38 USD
101786-462 101786-464
2-Fluorophenylhydrazinium chloride ≥97%
2-Fluorophenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012927
CAS Number: 2924-15-4
EINECS: 220-885-9

Order Now

Specification Test Results

Melting point 205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR