4-Fluorophenylhydrazinium chloride ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

003923-25G 003923-100G
101786-458EA 21.54 USD
101786-458 101786-460
4-Fluorophenylhydrazinium chloride ≥97% for synthesis
4-Fluorophenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₆H₈ClFN₂
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00012942
CAS Number: 823-85-8
EINECS: 212-521-2

Order Now

Specification Test Results

Melting point 250°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR