3-Formylphenylboronic acid ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid

003916-5G 003916-25G
101786-446EA 12.31 USD
101786-446 101786-448
3-Formylphenylboronic acid ≥97% for synthesis
3-Formylphenylboronic acid

Hazard code(s): Irritant, Moisture Sensitive, Store in Freezer

Formula: C₇H₇BO₃
Melting Pt: 181…183 °C
MDL Number: MFCD00161356
CAS Number: 87199-16-4

Order Now

Specification Test Results

Melting point 181-183°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR