4-Chlorophenylboronic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid

003914-25G 003914-100G
101786-440EA 36.92 USD
101786-440 102550-834
4-Chlorophenylboronic acid ≥98%
4-Chlorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 284…289 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Specification Test Results

Melting point 262-264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR