3-Chlorophenylboronic acid ≥97% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane
003913-25G
003913-100G
101786-436EA
70.77
USD
101786-436
101786-438
3-Chlorophenylboronic acid ≥97% for synthesis
3-Chlorophenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₆H₆BClO₂ Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161354 CAS Number: 63503-60-6 |
Specification Test Results
Melting point | 185-189°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...