2-Chlorophenylboronic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
003912-5G
003912-25G
101786-432EA
58.46
USD
101786-432
101786-434
2-Chlorophenylboronic acid ≥97%
2-Chlorophenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₆H₆BClO₂ MW: 156.38 g/mol Melting Pt: 176…179 °C |
MDL Number:
MFCD00674012 CAS Number: 3900-89-8 |
Specification Test Results
Melting point | 176-179°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...