3-Fluorophenylboronic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

003909-5G 003909-25G 003909-100G
101786-418EA 13.85 USD
101786-418 101786-420 101786-422
3-Fluorophenylboronic acid ≥97% for synthesis
3-Fluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BFO₂
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00236042
CAS Number: 768-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR