4-Fluorophenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

003907-25G 003907-100G
101786-412EA 29.23 USD
101786-412 101786-414
4-Fluorophenylboronic acid ≥97%
4-Fluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BFO₂
Melting Pt: 267…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00039136
CAS Number: 1765-93-1

Order Now

Specification Test Results

Melting point 267-268°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR