(4-Chlorophenyl)acetone ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Chlorophenylacetone
003905-5G
003905-25G
003905-100G
101786-406EA
21.54
USD
101786-406
101786-408
101786-410
(4-Chlorophenyl)acetone ≥97%
(4-Chlorophenyl)acetone
Hazard code(s): Irritant
Formula:
C₉H₉ClO MW: 168.62 g/mol Boiling Pt: 98 °C (3mm) Density: 1.13 g/cm³ |
MDL Number:
MFCD00045214 CAS Number: 5586-88-9 EINECS: 226-986-4 |
Specification Test Results
Boiling point | 98°C/3mm |
Density | 1.13 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...