(2-Ethoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

003891-1G 003891-5G
101786-376EA 36.92 USD
101786-376 101786-378
(2-Ethoxyphenyl)acetic acid ≥98%
(2-Ethoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: H₃CCH₂O(C₆H₄)CH₂COOH
Melting Pt: 103…104 °C
MDL Number: MFCD00016824
CAS Number: 70289-12-2

Order Now

Specification Test Results

Melting point 103-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR