(2-Fluorophenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid

003889-100G 003889-250G
101786-368EA 52.31 USD
101786-368 101786-370
(2-Fluorophenyl)acetic acid ≥98% for synthesis
(2-Fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄F)CH₂COOH
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 64 °C
MDL Number: MFCD00004315
CAS Number: 451-82-1
EINECS: 207-196-9

Order Now

Specification Test Results

Melting point 62-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR