3-Fluorophenyl acetate ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Acetoxy-3-fluorobenzene

003865-5G 003865-25G
101786-334EA 50.77 USD
101786-334 101786-336
3-Fluorophenyl acetate ≥98% for synthesis
3-Fluorophenyl acetate

Hazard code(s): Irritant

Formula: C₈H₇FO₂
Boiling Pt: 77…78 °C (13mm)
Density: 1.15 g/cm³
MDL Number: MFCD00142578
CAS Number: 701-83-7

Order Now

Specification Test Results

Boiling point 77-78°C/13m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR