4-Fluorophenetole ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Fluorophenyl ethyl ether, 1-Fluoro-4-ethoxybenzene, 1-Ethoxy-4-fluorobenzene
003860-1G
003860-5G
101786-330EA
40
USD
101786-330
101786-332
4-Fluorophenetole ≥98%
4-Fluorophenetole
Hazard code(s): Irritant
Formula:
C₈H₉FO MW: 140.16 g/mol |
MDL Number:
MFCD00017953 CAS Number: 459-26-7 EINECS: 207-287-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...