2-Fluorophenetole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Ethoxy-2-fluorobenzene, 2-Fluorophenyl ethyl ether, 1-Fluoro-2-ethoxybenzene

003859-1G 003859-5G 003859-25G
101786-324EA 21.54 USD
101786-324 101786-326 101786-328
2-Fluorophenetole ≥98%
2-Fluorophenetole

Hazard code(s): Irritant

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD00017909
CAS Number: 451-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR