4-Methylphenethylamine ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(p-Tolyl)ethylamine
003858-1G
003858-5G
101786-320EA
30.77
USD
101786-320
101786-322
4-Methylphenethylamine ≥98%
4-Methylphenethylamine
Hazard code(s): Irritant
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 214 °C Density: 0.93 g/cm³ |
MDL Number:
MFCD00008195 CAS Number: 3261-62-9 EINECS: 221-865-2 |
Specification Test Results
Boiling point | 214°C |
Density | 0.93 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...