2-(4-Fluorophenyl)ethanol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluorophenethyl alcohol

003852-5G 003852-25G 003852-100G
101786-310EA 49.23 USD
101786-310 101786-312 101786-314
2-(4-Fluorophenyl)ethanol ≥97%
2-(4-Fluorophenyl)ethanol

Hazard code(s): Irritant

Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 110…117 °C (20mm)
Density: 1.137 g/cm³ (20 °C)
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5

Order Now

Specification Test Results

Boiling point 110-117°C/2
Density 1.121
ND20 1.507

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR