1H,1H-Heneicosafluoro-1-undecanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heneicosafluoro-1-undecanol, 1H,1H-Perfluoro-1-undecanol, 1H,1H-Perfluoroundecan-1-ol

003831-5G 003831-25G
101786-296EA 60 USD
101786-296 101786-298
1H,1H-Heneicosafluoro-1-undecanol ≥95%
1H,1H-Heneicosafluoro-1-undecanol

Hazard code(s): Irritant

Formula: C₁₁H₃F₂₁O
MW: 550.11 g/mol
MDL Number: MFCD00153269
CAS Number: 307-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR