Dimethyl-2-fluoroterephthalate ≥97%

Supplier: Matrix Scientific

003826-1G 003826-5G
101786-292EA 118.46 USD
101786-292 101786-294
Dimethyl-2-fluoroterephthalate ≥97%
Dimethyl-2-fluoroterephthalate

Hazard code(s): Irritant

Formula: C₁₀H₉FO₄
Melting Pt: 84…86 °C
MDL Number: MFCD00042266
CAS Number: 5292-47-7
EINECS: 226-135-7

Order Now

Specification Test Results

Melting point 84-86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR