2,4,6-Trifluoropyrimidine ≥98% for synthesis

Supplier: Matrix Scientific
Danger

003818-1G 003818-5G
101786-286EA 66.15 USD
101786-286 101786-288
2,4,6-Trifluoropyrimidine ≥98% for synthesis
2,4,6-Trifluoropyrimidine

Hazard code(s): Corrosive, Moisture Sensitive, Stored under Argon

Formula: C₄HF₃N₂
Boiling Pt: 59 - 60 °C (1013 hPa)
MDL Number: MFCD00039705
CAS Number: 696-82-2
EINECS: 211-801-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 100-102°C
ND20 1.405


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR