2,3,4-Trifluorophenol ≥97%

Supplier: Matrix Scientific

003809-5G 003809-25G
101786-276EA 60 USD
101786-276 101786-278
2,3,4-Trifluorophenol ≥97%
2,3,4-Trifluorophenol

Hazard code(s): Irritant

Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 69 °C (43mm)
Melting Pt: 30…34 °C
Density: 1.46 g/cm³ (20 °C)
MDL Number: MFCD00042426
CAS Number: 2822-41-5
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Boiling point 69°C/43mm
Melting point 30-34°C
Density 1.46
ND20 1.465

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR