Bis(pentafluorophenyl)carbonate ≥97%

Supplier: Matrix Scientific

003806-5G 003806-25G
101786-268EA 58.46 USD
101786-268 101786-270
Bis(pentafluorophenyl)carbonate ≥97%
Bis(pentafluorophenyl)carbonate

Hazard code(s): Irritant

Formula: C₁₃F₁₀O₃
Melting Pt: 48…50 °C
MDL Number: MFCD00368353
CAS Number: 59483-84-0

Order Now

Specification Test Results

Melting point 48-50°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR