2-(2,4-Difluorophenoxy)-3-nitropyridine

Supplier: Matrix Scientific

003799-1G 003799-5G
101786-260EA 73.85 USD
101786-260 101786-262
2-(2,4-Difluorophenoxy)-3-nitropyridine
2-(2,4-Difluorophenoxy)-3-nitropyridine

Hazard code(s): Irritant

Formula: C₁₁H₆F₂N₂O₃
Melting Pt: 63…65 °C
MDL Number: MFCD00067800
CAS Number: 175135-62-3

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR