(3,4,5-Trifluorophenyl)acetic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
3,4,5-Trifluorophenylacetic acid
003790-1G
003790-5G
101786-254EA
29.23
USD
101786-254
101786-256
(3,4,5-Trifluorophenyl)acetic acid ≥97%
(3,4,5-Trifluorophenyl)acetic acid
Hazard code(s): Irritant
Formula:
(C₆H₂F₃)CH₂COOH MW: 190.12 g/mol |
MDL Number:
MFCD00083532 CAS Number: 209991-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...