(2,4,6-Trifluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4,6-Trifluorophenylacetic acid

003789-1G 003789-5G 003789-25G
101786-248EA 18.46 USD
101786-248 101786-250 101786-252
(2,4,6-Trifluorophenyl)acetic acid ≥97%
(2,4,6-Trifluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₂F₃)CH₂COOH
MW: 190.12 g/mol
MDL Number: MFCD00061218
CAS Number: 209991-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR