(2,3,6-Trifluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

003787-1G 003787-5G
101786-240EA 83.08 USD
101786-240 101786-242
(2,3,6-Trifluorophenyl)acetic acid ≥97%
(2,3,6-Trifluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: C₆H₂F₃CH₂COOH
Melting Pt: 111…114 °C
MDL Number: MFCD00061217
CAS Number: 114152-23-7

Order Now

Specification Test Results

Melting point 111-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR