(2,3,4-Trifluorophenyl)acetic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-(2,3,4-Trifluorophenyl)acetic acid
003785-1G
003785-5G
101786-236EA
32.31
USD
101786-236
101786-238
(2,3,4-Trifluorophenyl)acetic acid ≥97%
(2,3,4-Trifluorophenyl)acetic acid
Hazard code(s): Irritant
Formula:
C₆H₂F₃CH₂COOH MW: 190.12 g/mol |
MDL Number:
MFCD00061235 CAS Number: 243666-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...