2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H-Nonafluoro-1-pentanol, 1H,1H-Perfluoropentan-1-ol

003776-1G 003776-5G
101786-206EA 63.08 USD
101786-206 101786-208
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol ≥97%
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol

Hazard code(s): Irritant

Formula: C₅H₃F₉O
MW: 250.06 g/mol
Boiling Pt: 77 °C (173mm)
Density: 1.664 g/cm³
MDL Number: MFCD00236714
CAS Number: 355-28-2

Order Now

Specification Test Results

Boiling point 77°C/173mm
Density 1.664
ND20 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR