2H,3H-Perfluoropentane
Supplier: Matrix Scientific
|
Warning
|
003774-100G
003774-500G
101786-200EA
112.31
USD
101786-200
101786-202
2H,3H-Perfluoropentane
2H,3H-Perfluoropentane
Hazard code(s): Irritant, Refrigerate
Formula:
C₅H₂F₁₀ Boiling Pt: 53.6 °C Melting Pt: –80 °C Density: 1.580 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00798129 CAS Number: 138495-42-8 |
Specification Test Results
Boiling point | 53.6°C |
Melting point | -80°C |
Density | 1.58 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...