1,2,4-Trifluoro-5-nitrobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
2,4,5-Trifluoronitrobenzene
003761-25G
003761-100G
101786-192EA
26.15
USD
101786-192
101786-194
1,2,4-Trifluoro-5-nitrobenzene ≥98%
1,2,4-Trifluoro-5-nitrobenzene
Hazard code(s): Irritant, Lachrymator
Formula:
C₆H₂F₃NO₂ MW: 177.08 g/mol Boiling Pt: 81…82 °C (30mm) Melting Pt: –11 °C Density: 1.54 g/cm³ (20 °C) |
MDL Number:
MFCD00007051 CAS Number: 2105-61-5 EINECS: 218-281-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 81-82°C/30m |
Melting point | -11°C |
Density | 1.544 |
ND20 | 1.4938 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...