1,3,5-Trifluoro-2-nitrobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Nitro-2,4,6-trifluorobenzene, 2,4,6-Trifluoronitrobenzene
003760-25G
003760-100G
101786-188EA
146.15
USD
101786-188
101786-190
1,3,5-Trifluoro-2-nitrobenzene ≥98%
1,3,5-Trifluoro-2-nitrobenzene
Hazard code(s): Toxic
Formula:
C₆H₂F₃NO₂ MW: 177.08 g/mol Boiling Pt: 172 °C Melting Pt: 3.5 °C Density: 1.514 g/cm³ (20 °C) |
MDL Number:
MFCD00014687 CAS Number: 315-14-0 EINECS: 206-248-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 172°C |
Melting point | 3.5°C |
Density | 1.514 |
ND20 | 1.477 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...