1-Amino-2,2,2-trifluoroethane hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoroethanamine hydrochloride, 2,2,2-Trifluoroethylamine hydrochloride

003703-25G 003703-100G
101786-096EA 73.85 USD
101786-096 101786-098
1-Amino-2,2,2-trifluoroethane hydrochloride ≥98%
1-Amino-2,2,2-trifluoroethane hydrochloride

Hazard code(s): Irritant

Formula: C₂H₅ClF₃N
MW: 135.52 g/mol
Melting Pt: 220…222 °C
MDL Number: MFCD00012875
CAS Number: 373-88-6
EINECS: 206-771-1

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR