3,4,5-Trifluorocinnamic acid

Supplier: Matrix Scientific
Warning

003692-1G 003692-5G
101786-088EA 36.92 USD
101786-088 101786-090
3,4,5-Trifluorocinnamic acid
3,4,5-Trifluorocinnamic acid
Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
MDL Number: MFCD00083529
CAS Number: 152152-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR