2,3,4-Trifluorocinnamic acid

Supplier: Matrix Scientific
Warning

Synonyms: 3-(2,3,4-Trifluorophenyl)acrylic acid

003688-1G 003688-5G
101786-072EA 52.31 USD
101786-072 101786-074
2,3,4-Trifluorocinnamic acid
2,3,4-Trifluorocinnamic acid
Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
MDL Number: MFCD00083552
CAS Number: 207742-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR