Methyl-4-fluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 4-fluorobenzoate, 4-Fluorobenzoic acid emthyl ester

003680-25G 003680-100G
101786-068EA 21.54 USD
101786-068 101786-070
Methyl-4-fluorobenzoate ≥97%
Methyl-4-fluorobenzoate

Hazard code(s): Irritant

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 90…92 °C (20mm)
Density: 1.192 g/cm³
MDL Number: MFCD00017959
CAS Number: 403-33-8
EINECS: 206-956-7

Order Now

Specification Test Results

Boiling point 90-92°C/20m
Density 1.192
ND20 1.495

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR