2,3,4-Trifluorobenzaldehyde ≥98%
Supplier: Matrix Scientific
|
Warning
|
003677-1G
003677-5G
003677-25G
101786-062EA
12.31
USD
101786-062
101786-064
101786-066
2,3,4-Trifluorobenzaldehyde ≥98%
2,3,4-Trifluorobenzaldehyde
Hazard code(s): Irritant
Formula:
C₇H₃F₃O MW: 160.1 g/mol Boiling Pt: 77 - 79 °C (20 -rr) Density: 1.404 g/cm³ (20 °C) |
MDL Number:
MFCD00061230 CAS Number: 161793-17-5 |
Specification Test Results
Boiling point | 77-79°C/20m |
Density | 1.404 |
ND20 | 1.482 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...