2,3,6-Trifluorobenzaldehyde ≥97%

Supplier: Matrix Scientific

003672-1G 003672-5G 003672-25G
101786-052EA 18.46 USD
101786-052 101786-054 101786-056
2,3,6-Trifluorobenzaldehyde ≥97%
2,3,6-Trifluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 159 °C
Density: 1.425 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00061195
CAS Number: 104451-70-9

Order Now

Specification Test Results

Boiling point 159°C
Density 1.425
ND20 1.487

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR