2,3,6-Trifluorobenzylamine ≥97%

Supplier: Matrix Scientific
Danger

003659-1G 003659-5G
101786-034EA 52.31 USD
101786-034 101786-036
2,3,6-Trifluorobenzylamine ≥97%
2,3,6-Trifluorobenzylamine

Hazard code(s): Corrosive, Air Sensitive

Formula: C₇H₆F₃N
MW: 161.13 g/mol
MDL Number: MFCD00236315
CAS Number: 230295-09-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR