2,3,5-Trifluorobenzylamine
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
(2,3,5-Trifluorophenyl)methanamine
003658-1G
003658-5G
101786-030EA
187.69
USD
101786-030
101786-032
2,3,5-Trifluorobenzylamine
2,3,5-Trifluorobenzylamine
Hazard code(s): Irritant
Formula:
C₇H₆F₃N MW: 161.13 g/mol Boiling Pt: 73 °C (3mm) |
MDL Number:
MFCD00236314 CAS Number: 244022-72-8 UN: 2735 ADR: 8,III |
Specification Test Results
Boiling point | 73°C/3mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...