2,3,5-Trifluorobenzyl alcohol

Supplier: Matrix Scientific
Warning

003656-1G 003656-5G
101786-024EA 44.62 USD
101786-024 101786-026
2,3,5-Trifluorobenzyl alcohol
2,3,5-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
MDL Number: MFCD00083542
CAS Number: 67640-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR