2,3,5-Trifluorobenzamide

Supplier: Matrix Scientific

003653-1G 003653-5G
101786-020EA 36.92 USD
101786-020 101786-022
2,3,5-Trifluorobenzamide
2,3,5-Trifluorobenzamide
Formula: C₇H₄F₃NO
MDL Number: MFCD00061198
CAS Number: 238403-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR