2,3,5-Trifluorobenzaldehyde
Supplier: Matrix Scientific
|
Warning
|
003652-1G
003652-5G
101786-016EA
26.15
USD
101786-016
101786-018
2,3,5-Trifluorobenzaldehyde
2,3,5-Trifluorobenzaldehyde
Formula:
C₇H₃F₃O MW: 160.1 g/mol Boiling Pt: 165 °C (1013 hPa) Density: 1.407 g/cm³ (20 °C) |
MDL Number:
MFCD00083538 CAS Number: 126202-23-1 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...