3,4,5-Trifluorobenzyl alcohol ≥97%

Supplier: Matrix Scientific

003646-1G 003646-5G
101786-012EA 38.46 USD
101786-012 101786-014
3,4,5-Trifluorobenzyl alcohol ≥97%
3,4,5-Trifluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 203…204 °C
Density: 1.385 g/cm³
MDL Number: MFCD00083527
CAS Number: 220227-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR