2,4,6-Trifluorobenzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific
Warning

003645-1G 003645-5G
101786-008EA 18.46 USD
101786-008 101786-010
2,4,6-Trifluorobenzyl alcohol ≥97% for synthesis
2,4,6-Trifluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₅F₃O
MDL Number: MFCD00061207
CAS Number: 118289-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR